missing translation for 'onlineSavingsMsg'
Learn More

Smart-Vue™ Receivers

Choose from an assortment of Thermo Scientific™ Smart-Vue™ receivers including receiver packages that include power supply and software for your wireless monitoring solution.

Brand:  Thermo Scientific SV103-512-LSB

Additional Details : Weight : 0.37000kg

 View more versions of this product

Product Code. 15370321

  • 22128.89 NOK / Each


Currently not in stock
Add to basket

Specifications

Specifications

USB 1.0 or 2.0 Receiver, 25mW+SW 49 sensors
Receiver, 868MHz
868MHz
Velcro, CD ROM with Smart-Vue client server software 2.0 license up to 49 sensors, software drivers and user manuals.
Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Smart-Vue™ Wireless Monitoring Solutions
Videos