Learn More
Thermo Scientific™ Smart-Vue Pro Accessories
Customize your Smart-Vue Pro remote monitoring system with accessories to meet your specific needs.
Brand:  Thermo Scientific™ SVPACCCBL002
Additional Details : Weight : 0.13000kg
 View more versions of this product
Product Code. 16600076
  • 271.36 NOK / Each
Estimated Shipment: 21-12-2021
Add to basket
Description
Description
Choose from an assorment of Smart-Vue Pro accessories including power supplies, cables and mounting hardware, antenna and audible alarms to customize your remote monitoring system as well as ensure system integrity.
Compatible with Smart-Vue Pro remote monitoring system.
Specifications
Specifications
Smart-Vue Pro Duo Module, Smart-Vue Pro Quatro Module
Smart-Vue Pro supports conformance to 21 CFR Part 11, cGxP and SOP compliance
Smart-Vue Pro TCP/IP LAN Cable (CAT5) Length 2 m
Smart-Vue Pro Remote Monitoring System