missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad3 Antibody (378611) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB4038JF635
This item is not returnable.
View return policy
Description
Smad3 Monoclonal antibody specifically detects Smad3 in Human samples. It is validated for Immunohistochemistry,Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| Smad3 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| DKFZp586N0721, DKFZp686J10186, hMAD-3, hSMAD3, HsT17436, JV15-2MGC60396, MAD homolog 3, mad protein homolog, MAD, mothers against decapentaplegic homolog 3, MAD, mothers against decapentaplegic homolog 3 (Drosophila), mad3, MADH3mad homolog JV15-2, mothers against decapentaplegic homolog 3, Mothers against DPP homolog 3, SMA- and MAD-related protein 3, SMAD 3, SMAD family member 3HSPC193, SMAD, mothers against DPP homolog 3, SMAD, mothers against DPP homolog 3 (Drosophila), Smad3 | |
| E. coli-derived recombinant human Smad3, Ser2-Ala230, Accession # P84022 | |
| 0.1 mL | |
| Phospho Specific, Signal Transduction, Wnt Signaling Pathway | |
| 4088 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 378611 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction