missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SKIV2L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | SKIV2L2 |
---|---|
Concentration | 0.21 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890594
|
Proteintech
12719-2-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16880594
|
Proteintech
12719-2-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SKIV2L2 gene ontology annotations related to this gene include RNA catabolic process.Specifications
SKIV2L2 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P42285, Q9CZU3 | |
23517, 365668, 72198 | |
SKIV2L2 Fusion Protein Ag3414 | |
Primary | |
-20°C | |
SKIV2L2 |
0.21 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ATP dependent helicase SKIV2L2, Dob1, fSAP118, KIAA0052, Mtr4, SKIV2L2 | |
Skiv2l2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |