missing translation for 'onlineSavingsMsg'
Learn More

SIPA1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 26793-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16845155

  • 1415.13 NOK / 20µL
Estimated Shipment: 13-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The product of this gene is a mitogen induced GTPase activating protein. It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localized to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterized to date.
Specifications

Specifications

SIPA1
Polyclonal
Unconjugated
SIPA1
p130 SPA 1, Sipa 1, SIPA1, SPA1
Rabbit
Antigen Affinity Chromatography
RUO
6494
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.57 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q96FS4
SIPA1
SIPA1 Fusion Protein Ag25383
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers