missing translation for 'onlineSavingsMsg'
Learn More

PTPN2 Recombinant Mouse Protein, N-His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the amino acid (Met 1-Asn 314) of human PTPN2 (P17706-1) fused to the N-terminal polyhistidine-tagged GST tag at the N-terminus

Brand:  Invitrogen 50800M07B25

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15369266

  • 1286.00 NOK / 25µg


Estimated Shipment date
from Supplier 21-01-2019
Add to basket

Description

Description

Protein Tyrosine Phosphatase Nonreceptor Type 2 (PTPN2), recombinant mouse protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to dephosphorylate a phosphotyrosine residue in an EGF receptor (988-998 phosphopeptide substrate, R&D Systems Cat. No. ES006); the specific activity is >15nM/min/μg
  • Formulation: 20mM Tris, 500mM NaCl, 0.2μg/μL BSA, pH 8.0, 20% glycerol, 0.1mM TCEP
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
38.7kDa
5 x 5μg
Tyrosine Phosphatases
AI325124, Ptpt, TC-PTP
PTPN2
Insect
Frozen
≥85% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Protein tyrosine phosphatase, nonreceptor type 2
Assay Standard
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.