missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ IL1B Recombinant Canine Protein

This recombinant protein was expressed from a DNA sequence encoding the mature form of canine IL1B (Q28292) (Ala 115-Ser 266 ) fused to an initial Met at the N-terminus

Brand:  Thermo Scientific™ 70018DNAE5

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15369456

  • 862.00 NOK / 5µg


Estimated Shipment date
from Supplier 18-10-2018
Add to basket

Includes: One vial of lyophilized protein

Description

Description

Interleukin 1 Beta (IL1B), recombinant canine protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind human IL1R1 in a functional ELISA
  • Formulation: Lyophilized in 20mM PB, 0.3M NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
17.6kDa
5μg
Interleukins
Interleukin 1 beta
Assay Standard
E. coli
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
IL-1
IL1B
Cytokines & Receptors
Untagged
Videos

For Research Use Only. Not for use in diagnostic procedures.