Learn More
IL18 Recombinant Cynomolgus Protein, N-His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the Cynomolgus (Macaca fascicularis) IL-18 (Ala 2-Asp 193) fused to an N-terminal polyhistidine tag
Brand: Invitrogen 90011C07E25
Additional Details : Weight : 0.10000kg
Includes: Five vials of lyophilized protein
Description
Interleukin 18 (IL18), recombinant cynomolgus protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: Measured by its ability to bind human IL18R1 in a functional ELISA
- Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
5 x 5μg | |
IL-18 | |
IL18 | |
E. coli | |
Lyophilized | |
≥90% by SDS-PAGE |
23.7kDa | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed. | |
Interleukin 18 | |
His Tag | |
Cytokines & Receptors | |
Interleukins |
For Research Use Only. Not for use in diagnostic procedures.