missing translation for 'onlineSavingsMsg'
Learn More

IDO1 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human IDO1 (P14902-1) (Ala 2-Gly 403) fused to a polyhistidine tag at the N-terminus

Brand:  Invitrogen 11650H07E25

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15208785

  • 4185.00 NOK / 25µg


Estimated Shipment
20-08-2020
Add to basket

Description

Description

Indoleamine 2,3-Dioxygenase 1 (IDO1), recombinant human protein is supplied as a frozen protein. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 50mM Tris, 150mM NaCl, 10% Glycerol, pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

E. coli
Assay Standard
Frozen
46kDa
≥85% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
IDO, INDO
IDO1
Other Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3620
PAGE
5 x 5μg
Full Length/Recombinant
Indoleamine 2,3-Dioxygenase 1
His Tag
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.