missing translation for 'onlineSavingsMsg'
Learn More

GPNMB Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human GPNMB isoform 2 (Q14956-2) extracellular domain (Met 1-Pro 474) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 11305H08H5

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15208515

  • 580.00 NOK / 5µg


Estimated Shipment
19-08-2020
Add to basket

Description

Description

Glycoprotein (Transmembrane)nMB (GPNMB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Other Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
10457
PAGE
5μg
C-Terminal
<1 EU/μg
Glycoprotein (transmembrane)nMb
His Tag
Other Proteins
Assay Standard
Lyophilized
52.1kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
UNQ1725/PRO9925, HGFIN,nMB
GPNMB
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.