missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ EREG Recombinant Mouse Protein

This recombinant protein was expressed from a DNA sequence encoding the mature form of mouse EREG (Q61521) (Val 56-Leu 101)

Brand:  Thermo Scientific™ 50599M01H5

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15369026

  • 586.00 NOK / 5µg


Estimated Shipment date
from Supplier 25-06-2018
Add to basket

Includes: One vial of lyophilized protein

Description

Description

Epiregulin (EREG), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
33.8kDa
5μg
EGF (Epidermal Growth Factor)|Growth Factor Receptors
EPR, mgC36144
EREG
Growth Factors & Receptors
Untagged
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Proepiregulin
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.