missing translation for 'onlineSavingsMsg'
Learn More

DLL4 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse DLL4 (NP_062327.2) (Met 1-Pro 525) fused to a polyhistidine tag at the C-terminus

Brand:  Invitrogen 50640M08H25

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15430624

  • 1280.00 NOK / 25µg


Estimated Shipment
20-08-2020
Add to basket

Includes: Five vials of lyophilized protein

Description

Description

Delta-like 4 (Drosophila,) (DLL4), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized mouse DLL4-His at 10μg/mL (100μL/well) can bind biotinylated mouse NOTCH1-His; the EC50 of biotinylated mouse NOTCH1-His is 30ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Neurotrophic Factors
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
55.7kDa
5 x 5μg
<1 EU/μg
Delta-like 4 (Drosophila)
His Tag
Other Proteins
Assay Standard
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Dll4, RP23-46P4.8, Delta4
DLL4
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.