missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Silver(I) oxide, 99.99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Polishing glass, coloring glass yellow, catalyst, purifying drinking water, lab reagent, carbon dioxide scrubber, and chemical sensors. It is used in the preparation of other silver compounds, and silver-oxide batteries. In organic chemistry, silver oxide finds use as an oxidizing agent for aldehyes to produce carboxylic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 230°C (decomposition) |
Color | Brown |
Physical Form | Powder |
Quantity | 1 g |
Assay Percent Range | 99.99% (metals basis) |
Solubility Information | Soluble in dilute nitric acid and ammonia. Practically insoluble in water and alcohol |
Formula Weight | 231.74 |
Odor | Odorless |
Sensitivity | Light sensitive |
Appearance | Dark brown powder |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'