missing translation for 'onlineSavingsMsg'
Learn More

Silver cyanate, 98+%, Thermo Scientific Chemicals

Catalog Number 10563391
Click to view available options
Quantity:
10 g
25 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3315-16-0
CAgNO
149.89
VAQVDZZAAMAZAO-UHFFFAOYSA-N
silver,cyanate, cyanic acid; silver
45051745
cyanic acid;silver
C(#N)O.[Ag]
This item is not returnable. View return policy
3315-16-0
CAgNO
149.89
VAQVDZZAAMAZAO-UHFFFAOYSA-N
silver,cyanate, cyanic acid; silver
45051745
cyanic acid;silver
C(#N)O.[Ag]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1085.0°C
Melting Point 652.0°C
Color Beige to Gray
Physical Form Powder
Quantity 50 g
Assay Percent Range 98% min
Linear Formula AgOCN
Fieser 02,224; 03,162
Solubility Information Solubility in water: soluble
Formula Weight 149.89
Percent Purity ≥98%
Packaging Glass bottle
Chemical Name or Material Silver cyanate
Show More Show Less
compliance-icons
Product Identifier
  • Silver cyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'