missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ Silicone Pump Tubing

Brand:  Velp Scientifica™ 10002042

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 15369514

  • 90.85 NOK / Each


Estimated Shipment
07-01-2019
Add to basket

Specifications

Specifications

7mm
2 to 35mL/min.
Pump Tubing
Transfer of Liquids, Gases, Mixtures, Emulsions, etc
1Each
4mm
Silicone
100cm
SP 311/2 Peristaltic Pump
4mm I.D./7mm O.D.
Videos