Learn More
Ismatec™ Peroxide-Cured Silicone Pump Tubing
Reduce the risk of particulate entrapment with smooth inner surface
Brand: Ismatec™ 07625-42
Additional Details : Weight : 0.01000kg
Description
- Tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Regulatory approvals to match your unique application
Specifications
Extension Tubing | |
2.06 mm | |
15 m | |
-50°C | |
42 |
Yes | |
Silicone | |
C/L (Microbore/Autoanalysis) Pump Series | |
1 Pack | |
230°C |