Learn More
Cole-Parmer™ Silicone 210 Bath Fluid
Brand:  Cole-Parmer™ 01294-60
Additional Details : Weight : 4.50000kg
Product Code. 11700399
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
-65°C to +280°C
Silicone 210 Bath Fluid
198 cp at 26°C