Learn More
BRAND™ SILBERBRAND Automatic Burette with Lateral Stopcock and Bottle
SILBERBRAND Automatic Burette with Lateral Stopcock and Bottle is constructed of amber borosilicate glass.
Brand: BRAND™ 22312
Additional Details : Weight : 1.96200kg
Description
- Borosilicate glass 5.4
- Error limits according to Class B, DIN EN ISO 385
- Interchangeable keys
- Calibrated to deliver (TD, Ex)
- With glass key in the titration stopcock
Specifications
Amber | |
0.1 mL | |
PTFE lateral stopcock | |
DIN EN ISO 385 |
25 mL | |
2000 mL bottle | |
12 months |