missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHMT1 Antibody [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-21429JF635
This item is not returnable.
View return policy
Description
SHMT1 Polyclonal antibody specifically detects SHMT1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| SHMT1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Janelia Fluor 635 | |
| 14 kDa protein, CSHMT, cytoplasmic serine hydroxymethyltransferase, Glycine hydroxymethyltransferase, MGC15229, MGC24556, serine hydroxymethyltransferase 1 (soluble), serine hydroxymethyltransferase, cytosolic, Serine methylase, SHMTEC 2.1.2.1 | |
| Produced in rabbits immunized with E. coli-derived Human SHMT1 fragment | |
| 0.1 mL | |
| metabolism | |
| 6470 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction