missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Stainless-Steel Instrument Trays, 8.9L x 5W x 2 in. D (22.5 x 12.7 x 5.1cm)
Epredia™ Shandon Stainless-Steel Instrument Trays are available in two sizes.
Brand: Epredia™ 73092
Additional Details : Weight : 2.00000kg
Description
- Constructed of surgical grade stainless steel
Specifications
Instrument Trays | |
8.9 x 5 x 2 in. (22.3 x 12.7 x 5.1 cm) | |
22.5 cm | |
12.7 cm | |
5.1 cm |
Stainless Steel | |
8.9 in. | |
5 in. | |
2 in. |