Learn More
SGE™ SilFlow™ GC 3 Port Splitter: Device Only
Highly efficient and reliable micro fluidic platform to improve GC connectivity
5590.00 NOK
Specifications
Type Port Splitter
For Use With SGE Silflow Microchannel Device
View More Specs
Products 3
Product Code Brand Diameter (Metric) Inner Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Diameter (Metric) Outer Price Quantity & Availability  
15299173
View Documents Promotion Details
SGE™
123723
Ports B and C 0.25/0.32 mm 1.1 mm
5590.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 21-05-2024
to see stock.
Add to basket
15209183
View Documents Promotion Details
SGE™
123724
Ports B and C 0.25/0.32 mm 0.53 mm
5590.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
15219183
View Documents Promotion Details
SGE™
123725
0.25/0.32 mm 0.25/0.32 mm
5590.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
Description
  • 1.1mm OD tubing not included
Specifications
Specifications
Port Splitter
SGE Silflow Microchannel Device