Learn More
Trajan™ Micro Control Valves
Precision metering and on-off control of gas flows. Designed specifically for chromatography applications and may be used at elevated temperatures from high vacuum to medium pressure.
2000.00 NOK - 7975.00 NOK
Specifications
Material | Stainless Steel |
---|---|
Connection Size (Metric) | 0.15 cm |
Max. Pressure | 700 psi |
Product Code | Brand | Valve Code | Length (Metric) Stem | Style | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Valve Code | Length (Metric) Stem | Style | Price | Quantity & Availability | |||
15289153
|
Trajan™
1236103 |
MOVPT | 50 mm | Pneumatic T Dual Outlet |
7975.00 NOK
Each |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15269153
|
Trajan™
1236091 |
MOVP | 50 mm | Pneumatic L Single Outlet |
7475.00 NOK
Each |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12689949
|
Trajan™
1236104 |
MOVPT(L) | 100 mm | Pneumatic T Dual Outlet |
7975.00 NOK
Each |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15209163
|
Trajan™
1236283 |
SMOV | L Single Outlet Manual |
2000.00 NOK
Each |
Estimated Shipment: 24-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Ideal for chromatography applications
- May be used at elevated temperatures from high vacuum to medium pressure
- Vacuum rated to a leak rate of 1E-05 cc/sec of helium
Specifications
Stainless Steel | |
0.15 cm | |
700 psi |