Learn More
Trajan™ Micro Control Valves
Precision metering and on-off control of gas flows. Designed specifically for chromatography applications and may be used at elevated temperatures from high vacuum to medium pressure.
Brand: Trajan™ 1236091
Additional Details : Weight : 0.01000kg
Description
- Ideal for chromatography applications
- May be used at elevated temperatures from high vacuum to medium pressure
- Vacuum rated to a leak rate of 1E-05 cc/sec of helium
Specifications
Pneumatic Valve | |
1/16 in. | |
0.15 cm | |
50 mm | |
700 psi | |
300°C |
Pneumatic L Single Outlet | |
Stainless Steel | |
1.9 in. | |
MOVP | |
572°F |