missing translation for 'onlineSavingsMsg'
Learn More

SF-1/NR5A1/Steroidogenic Factor 1 Antibody (SF1/8000R) [Janelia Fluor™ 585], Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-20854JF585

Product Code. 30171791

  • 6275.00 NOK / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

SF-1/NR5A1/Steroidogenic Factor 1 Monoclonal antibody specifically detects SF-1/NR5A1/Steroidogenic Factor 1 in Human,Rat samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SF-1/NR5A1/Steroidogenic Factor 1
Monoclonal
Janelia Fluor 585
AD4BPSTF-1, Adrenal 4-binding protein, ELP, FTZ1adrenal 4 binding protein, FTZF1nuclear receptor AdBP4, Fushi tarazu factor homolog 1, Nuclear receptor subfamily 5 group A member 1, nuclear receptor subfamily 5, group A, member 1, SF-1POF7, SF1steroidogenic factor 1, Steroid hormone receptor Ad4BP, steroidogenic factor-1
Recombinant fragment (around aa200-400) of human SF-1/NR5A1/Steroidogenic Factor 1 protein (exact sequence is proprietary)
0.1 mL
Cell Biology
2516
Store at 4°C in the dark.
IgG κ
Immunohistochemistry (Paraffin)
SF1/8000R
50mM Sodium Borate
Rabbit
Protein A or G purified
RUO
Primary
Human, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.