missing translation for 'onlineSavingsMsg'
Learn More

Serotonin hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7025650
Click to view available options
Quantity:
1 g
5 g
10 g
153-98-0
C10H13ClN2O
212.677
MFCD00012686
MDIGAZPGKJFIAH-UHFFFAOYSA-N
serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride
160436
3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride
C1=CC2=C(C=C1O)C(=CN2)CCN.Cl
153-98-0
C10H13ClN2O
212.677
MFCD00012686
MDIGAZPGKJFIAH-UHFFFAOYSA-N
serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride
160436
3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride
C1=CC2=C(C=C1O)C(=CN2)CCN.Cl

Serotonin hydrochloride is used as a neurotransmitter and function as an endogenous serotonin receptor agonist in various biochemical processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Serotonin Hydrochloride is used as a neurotransmitter and function as an endogenous serotonin receptor agonist in various biochemical processes.

Solubility
Soluble in water, alcohol, hydrochloric acid and dimethyl sulfoxide. Insoluble in ether and chloroform.

Notes
Store in a cool place. Light sensitive and hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 167°C to 172°C
Assay Percent Range 0.99
Quantity 1 g
UN Number UN2811
Beilstein 3570963
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,8463
Solubility Information Soluble in water,alcohol,hydrochloric acid and dimethyl sulfoxide. Insoluble in ether and chloroform.
Formula Weight 212.68
Percent Purity 98%
Chemical Name or Material Serotonin hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'