Learn More
SD-208, MedChemExpress
Product Code. 16455568
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16455568
Brand: Medchem Express HY13227
MedChemExpress SD-208 is a selective TGF-βRI (ALK5) inhibitor with IC50 of 48 nM, and > 100-fold selectivity over TGF-βRII.
SD-208 is a selective TGF-βRI (ALK5) inhibitor with IC50 of 48 nM, and > 100-fold selectivity over TGF-βRII.
Specifications
Chemical Name or Material SD-208
CAS 627536-09-8
Color Yellow-Brown
Purity Grade Notes Research
Molecular Formula C17H10ClFN6
Quantity 10 mg
Solubility Information DMSO : 9.09 mg/mL (25.77 mM; Need ultrasonic)
SMILES FC1=CC=C(Cl)C=C1C2=NC3=NC=CN=C3C(NC4=CC=NC=C4)=N2
Molecular Weight (g/mol) 352.75
Formula Weight 352.75
Percent Purity 99.42%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.