missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S1 protein (N501Y), His Tag
5799.57 NOK - 43038.87 NOK
Specifications
Accession Number | QHD43416.1 (N501Y) |
---|---|
Molecular Weight (g/mol) | 76.9 kDa |
Source | SARS-CoV-2 S1 protein (N501Y), His Tag (S1N-C52Hg) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1 (N501Y)). N501Y mutaion was identified in both the SARS-CoV-2 variant (known as 20 B/501 Y.V1, VOC 202012/01, or B0.1.1.7 lineage) which emerged in the United Kindom and SARS-CoV-2 variant (known as 20 C/501 Y.V2 or B0.1.351 lineage) which emerged in South Africa. |
Immunogen | Val 16 - Arg 685 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17206470
|
ACROBiosystems
S1N-C52HG-100UG |
100 μg |
5799.57 NOK
100µg |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17216470
|
ACROBiosystems
S1N-C52HG-1MG |
1 mg |
43038.87 NOK
1mg |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.9 kDa. The protein migrates as 110-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
QHD43416.1 (N501Y) | |
SARS-CoV-2 S1 protein (N501Y), His Tag (S1N-C52Hg) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1 (N501Y)). N501Y mutaion was identified in both the SARS-CoV-2 variant (known as 20 B/501 Y.V1, VOC 202012/01, or B0.1.1.7 lineage) which emerged in the United Kindom and SARS-CoV-2 variant (known as 20 C/501 Y.V2 or B0.1.351 lineage) which emerged in South Africa. | |
-20°C | |
S1 protein | |
SARS-CoV-2 | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |
76.9 kDa | |
Val 16 - Arg 685 | |
Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1,COVID-19 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |