Learn More
Cole-Parmer™ Acid Washed Grinding Beads
Acid Washed Grinding Beads are treated to remove fine particles and contaminants.
2560.00 NOK - 3450.00 NOK
Specifications
Product Type | Grinding Beads |
---|---|
For Use With (Application) | Grinding |
For Use With (Equipment) | 2010 Geno/Grinder |
Packaging Quantity | 200 g |
Particle Size | 800 to 1000 μm |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
12879045
|
Cole-Parmer™
2160 |
No Grade |
2560.00 NOK
200g |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
12534166
|
Cole-Parmer™
2161 |
Molecular Biology Grade |
3450.00 NOK
200g |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||||
Description
- Choose from silica or zirconia grinding beads to help remove fine particles and contaminant.
- Using untreated beads which contain impurities can affect homogenization and biological processes.
- Acid washing silica and zirconia grinding beads help remove fine particles and contaminant for cleaner samples.
- Select silica beads for general grinding applications; zirconia beads have greater density and offer higher impact power.
- General-purpose grinding beads are suitable for most basic cell lysis applications.
- Choose RNase/DNase-free treated molecular biology-grade grinding beads to inactivate contaminating enzymes.
- Low-binding grinding beads are coated to reduce nonspecific binding of nucleic acids and proteins.
Specifications
Grinding Beads | |
2010 Geno/Grinder | |
800 to 1000 μm |
Grinding | |
200 g |