missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sam68 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10222-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This is a rabbit polyclonal antibody raised against residues near the N-terminus of human KHDRBS1.
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. Pseudogenes of this gene are found on chromosomes 5, 6, and 14. Two transcript variants encoding different isoforms have been found for this gene.Specifications
Sam68 | |
Polyclonal | |
Unconjugated | |
KHDRBS1 | |
KHDRBS1, p62, p68, Sam68 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10657, 117268, 20218 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q07666, Q60749 | |
KHDRBS1 | |
Sam68 Fusion Protein Ag0235 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |