missing translation for 'onlineSavingsMsg'
Learn More

Salicylic acid, For ACS analysis, 99.68%, MP Biomedicals™

Catalog Number p-4404949
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
69-72-7
C7H6O3
138.12
MFCD00002439
YGSDEFSMJLZEOE-UHFFFAOYSA-N
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
338
CHEBI:16914
2-hydroxybenzoic acid
OC(=O)C1=CC=CC=C1O
This item is not returnable. View return policy
69-72-7
C7H6O3
138.12
MFCD00002439
YGSDEFSMJLZEOE-UHFFFAOYSA-N
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
338
CHEBI:16914
2-hydroxybenzoic acid
OC(=O)C1=CC=CC=C1O
Color White
Assay Percent Range >99%
Formula Weight 138.12
Density 1.443g/mL
Quantity 100 g
Physical Form Powder
Chemical Name or Material Salicylic acid, For ACS analysis
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'