missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-Aminomethyl-1-Boc-pyrrolidine, 97%, Thermo Scientific Chemicals
927.64 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
Description
(S)-2-Aminomethyl-1-Boc-pyrrolidine acts as an intermediate in organic synthesis. It is also used as a chiral and heterocyclic building block in synthetic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(S)-2-Aminomethyl-1-Boc-pyrrolidine acts as an intermediate in organic synthesis. It is also used as a chiral and heterocyclic building block in synthetic chemistry.
Solubility
Miscible with N-methylpyrrolidinone.
Notes
Air and moisture sensitive. Store in a cool place. Incompatible with oxidizing agents, strong acids and bases.
(S)-2-Aminomethyl-1-Boc-pyrrolidine acts as an intermediate in organic synthesis. It is also used as a chiral and heterocyclic building block in synthetic chemistry.
Solubility
Miscible with N-methylpyrrolidinone.
Notes
Air and moisture sensitive. Store in a cool place. Incompatible with oxidizing agents, strong acids and bases.
Specifications
Boiling Point | 98°C to 112°C (1 mmHg) |
Quantity | 1 g |
UN Number | UN2735 |
Solubility Information | Miscible with N-methylpyrrolidinone. |
Formula Weight | 200.28 |
Percent Purity | 97% |
Chemical Name or Material | (S)-2-Aminomethyl-1-Boc-pyrrolidine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'