missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Methoxyphenyl)ethylamine, ChiPros 99+%, ee 98%, Thermo Scientific Chemicals

Catalog Number p-7046782
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
41851-59-6
C9H13NO
151.209
MFCD00671660
JTDGKQNNPKXKII-ZETCQYMHSA-N
s---1-4-methoxyphenyl ethylamine, s-1-4-methoxyphenyl ethanamine, s-1-4-methoxyphenyl ethylamine, s-1-4-methoxy-phenyl-ethylamine, 1s-1-4-methoxyphenyl ethanamine, s--1-4-methoxyphenyl ethylamine, s-p-1-4-methoxyphenyl ethylamine, s-1-4-methoxyphenyl ethan-1-amine, 1s-1-4-methoxyphenyl ethan-1-amine
793467
(1S)-1-(4-methoxyphenyl)ethanamine
CC(C1=CC=C(C=C1)OC)N
This item is not returnable. View return policy
41851-59-6
C9H13NO
151.209
MFCD00671660
JTDGKQNNPKXKII-ZETCQYMHSA-N
s---1-4-methoxyphenyl ethylamine, s-1-4-methoxyphenyl ethanamine, s-1-4-methoxyphenyl ethylamine, s-1-4-methoxy-phenyl-ethylamine, 1s-1-4-methoxyphenyl ethanamine, s--1-4-methoxyphenyl ethylamine, s-p-1-4-methoxyphenyl ethylamine, s-1-4-methoxyphenyl ethan-1-amine, 1s-1-4-methoxyphenyl ethan-1-amine
793467
(1S)-1-(4-methoxyphenyl)ethanamine
CC(C1=CC=C(C=C1)OC)N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point <-20°C
Density 1.02
Boiling Point 65°C (0.38 mmHg)
Refractive Index 1.533
Quantity 1 g
UN Number UN2735
Beilstein 3196456
Sensitivity Air Sensitive
Optical Rotation −34° (Neat)
Formula Weight 151.21
Percent Purity ≥99%
Assay ee 98%
Chemical Name or Material (S)-(-)-1-(4-Methoxyphenyl)ethylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'