missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Fluorophenyl)ethylamine, ChiPros 99%, ee 99%, Thermo Scientific Chemicals

Catalog Number p-7047404
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
66399-30-2
C8H10FN
139.173
MFCD03092999
QGCLEUGNYRXBMZ-LURJTMIESA-N
s-1-4-fluorophenyl ethylamine, s-1-4-fluorophenyl ethanamine, 1s-1-4-fluorophenyl ethanamine, 1s-1-4-fluorophenyl ethylamine, s-1-4-fluoro-phenyl-ethylamine, s---1-4-fluorophenyl ethylamine, 1s-1-4-fluorophenyl ethan-1-amine, pubchem15218, 5-1-4-fluorophenyl ethanamine, s-4-fluoro-, a-methylbenzylamine
2779063
(1S)-1-(4-fluorophenyl)ethanamine
CC(C1=CC=C(C=C1)F)N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
66399-30-2
C8H10FN
139.173
MFCD03092999
QGCLEUGNYRXBMZ-LURJTMIESA-N
s-1-4-fluorophenyl ethylamine, s-1-4-fluorophenyl ethanamine, 1s-1-4-fluorophenyl ethanamine, 1s-1-4-fluorophenyl ethylamine, s-1-4-fluoro-phenyl-ethylamine, s---1-4-fluorophenyl ethylamine, 1s-1-4-fluorophenyl ethan-1-amine, pubchem15218, 5-1-4-fluorophenyl ethanamine, s-4-fluoro-, a-methylbenzylamine
2779063
(1S)-1-(4-fluorophenyl)ethanamine
CC(C1=CC=C(C=C1)F)N

(S)-(-)-1-(4-Fluorophenyl)ethylamine is used in organic synthesis. It acts as a ligand and form a coordination complex, diacetato-2-O-bis[(S)-1-(4-fluorophenyl)ethylamine-N]palladium(II) by reacting with palladium(II) acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-(4-Fluorophenyl)ethylamine is used in organic synthesis. It acts as a ligand and form a coordination complex, diacetato-2-O-bis[(S)-1-(4-fluorophenyl)ethylamine-N]palladium(II) by reacting with palladium(II) acetate.

Solubility
Miscible with dimethyl sulfoxide.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -30°C
Density 1.03
Boiling Point 76°C (22 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.502
Quantity 1 g
UN Number UN2735
Beilstein 6791614
Sensitivity Air Sensitive
Solubility Information Miscible with dimethyl sulfoxide.
Optical Rotation −33° (Neat)
Formula Weight 139.17
Percent Purity 99%
Assay ee 99%
Chemical Name or Material (S)-(-)-1-(4-Fluorophenyl)ethylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'