Learn More
(S)-1,4-Diazabicyclo[4.3.0]nonane, 98%, ACROS Organics™
Brand:  Acros Organics 456810010
Additional Details : CAS Number : 93643-24-4 Weight : 0.00100kg Transport : UN number : 3267 Chem class : 8 Pack group : III
Quantity 1g
 View more versions of this product
Product Code. 15442182
  • 6053.00 NOK / 1g
Estimated Shipment
14-12-2020
Chemical Identifiers
93643-24-4
126.2
s-octahydropyrrolo 1,2-a pyrazine, s-1,4-diazabicyclo 4.3.0 nonane, 8as-octahydropyrrolo 1,2-a pyrazine, 8as-octahydropyrrolo 1,2-a piperazine, s-octahydro-pyrrolo 1,2-a pyrazine, 6s-1,4-diazabicyclo 4.3.0 nonane, pyrrolo 1,2-a pyrazine, octahydro-, 8as, 8as-1,2,3,4,6,7,8,8a-octahydropyrrolo 1,2-a pyrazine, pubchem20597, 6s-1,4-diazabicyclo 4,3,0 nonane
(8aS)-1,2,3,4,6,7,8,8a-octahydropyrrolo[1,2-a]pyrazine
C7H14N2
FTTATHOUSOIFOQ-ZETCQYMHSA-N
781249
C1CC2CNCCN2C1
Specifications
(S)-1,4-Diazabicyclo[4.3.0]nonane
Authentic
+ 5.88
93643-24-4
C7H14N2
FTTATHOUSOIFOQ-ZETCQYMHSA-N
(8aS)-1,2,3,4,6,7,8,8a-octahydropyrrolo[1,2-a]pyrazine
781249
98%
89°C
+ 5.88 (20.00°C c=1, CHCl3)
1g
97.5% min. (GC)
s-octahydropyrrolo 1,2-a pyrazine, s-1,4-diazabicyclo 4.3.0 nonane, 8as-octahydropyrrolo 1,2-a pyrazine, 8as-octahydropyrrolo 1,2-a piperazine, s-octahydro-pyrrolo 1,2-a pyrazine, 6s-1,4-diazabicyclo 4.3.0 nonane, pyrrolo 1,2-a pyrazine, octahydro-, 8as, 8as-1,2,3,4,6,7,8,8a-octahydropyrrolo 1,2-a pyrazine, pubchem20597, 6s-1,4-diazabicyclo 4,3,0 nonane
C1CC2CNCCN2C1
126.2
126.2
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • (S)-1,4-Diazabicyclo¢4.3.0!nonane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.