missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX3/CBFA3 Antibody (527327) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3765JF525
This item is not returnable.
View return policy
Description
RUNX3/CBFA3 Monoclonal antibody specifically detects RUNX3/CBFA3 in Human,Mouse samples. It is validated for Western Blot,Simple Western,Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| RUNX3/CBFA3 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Acute myeloid leukemia 2 protein, AML2SL3/AKV core-binding factor alpha C subunit, CBFA3MGC16070, CBF-alpha-3, PEA2 alpha C, PEA2-alpha C, PEBP2 alpha C, PEBP2A3FLJ34510, PEBP2-alpha C, runt domain, alpha subunit 3, runt-related transcription factor 3, SL3-3 enhancer factor 1 alpha C subunit, transcription factor AML2 | |
| E. coli-derived recombinant human RUNX3/CBFA3, Lys186-Tyr415, Accession # Q13761 | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 864 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 527327 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction