Learn More
Macherey Nagel X100 Round filter MN 651, D=32cm .
Brand:  Macherey Nagel 445032
X100 Round filter MN 651, D=32cm .
Product Code. 10012811
  • ONLINE EXCLUSIVE PRICE
    772.00 NOK
    568.27 NOK / Pack of 100
    Save 203.73 NOK
    26% Off
Estimated Shipment: 06-11-2026