missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RREB1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | RREB1 |
---|---|
Concentration | 333 μg/mL |
Applications | Western Blot, Immunohistochemistry, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16885775
|
Proteintech
27706-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16875775
|
Proteintech
27706-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The RREB1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RREB1. This antibody recognizes human antigen. The RREB1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
RREB1 | |
Western Blot, Immunohistochemistry, ELISA | |
Unconjugated | |
Human | |
IgG |
333 μg/mL | |
Polyclonal | |
Rabbit | |
6239 | |
Primary |