missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS3 Mouse anti-Human, Mouse, Rat, Clone: 2G7H4, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | RPS3 |
---|---|
Clone | 2G7H4 |
Concentration | 1.12 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887565
|
Proteintech
66046-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16877565
|
Proteintech
66046-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
WD repeat domain 5 (WDR5) is a member of the WD repeat protein family, which is involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. WDR5, also known as BIG-3, is expressed in the developing growth plate, accelerates chondrocyte and osteoblast differentiation in vitro, and regulates osteoblast differentiation during embryonic bone development. WDR5 interacts with the pluripotency factor Oct4/POU5F1 and is required for the efficient formation of induced pluripotent stem (iPS) cells.Specifications
RPS3 | |
1.12 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
40S ribosomal protein S3, ribosomal protein S3, RPS3 | |
RPS3 | |
IgG2a | |
Protein A | |
Antibody |
2G7H4 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
P23396, P62908, P62909 | |
140654, 27050, 6188 | |
RPS3 Fusion Protein Ag7515 | |
Primary | |
-20°C | |
RPS3 |