Learn More
Lonza™ RPMI 1640 with UltraGlutaminetm, 500mL
Developed for culturing normal human leukocytes. Lonza™ RPMI 1640 with UltraGlutaminetm, 500mL can be used for suspension and monolayer mammalian cultures.
Brand: Lonza™ BE12-702F
Additional Details : Weight : 0.64000kg
Description
- Developed for culturing normal human leukocytes
- Can be used for suspension and monolayer mammalian cultures
- Will work with different cell lines
- Can also be used with primary cells such as blood/hematopoietc cells
Specifications
Cell Culture Media | |
2 g/L D-Glucose anhydrous, 2 g/L NaHCO3, 300mg/L L-Glutamine, 400mg/L KCl, 6 g/L NaCl, 100mg/L Ca(NO3)2.4H2O, 1mg/L Glutathione reduce, 241.86mg/L L-Arginine. HCl, 56.82mg/L L-Asparagine.H2O, 20mg/L L-Aspartic Acid, 10mg/L Glycine, 20mg/L L-Glutamic Acid, | |
500 mL |