Learn More
Hettich™ Rotor Adapter for Carrier no. 1752
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
1225.00 NOK
Specifications
Type | Rotor Adapter |
---|---|
For Use With (Equipment) | ROTINA 380/380 R Centrifuge with Carrier no. 1752 |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
11494978
|
Hettich™
1763-A |
4 to 15 mL Tubes |
1225.00 NOK
Each |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11414988
|
Hettich™
1765 |
3 x 45 to 75 mL Tubes |
1225.00 NOK
Each |
Estimated Shipment: 27-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10556575
|
Hettich™
1761 |
18 x 3 mL Tubes or 36 x 1.5 to 2 mL Tubes |
1225.00 NOK
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
Description
- For use with ROTINA 380/380 R
- The ROTINA 380's versatility and size make an invaluable addition to any lab. Ideal for clinical, research, food and beverage and industrial applications.
- Robust and steady high-capacity centrifuge
- Accessories with bio-containment protect users from aerosols while ensuring optimal safety, for instance, while centrifuging infectious samples
- Temperature control system with pre-cool function from -20 to +40°C
- Versa-Rotor™ for multiple applications
- All-steel construction
- Quiet Operation
- Easy-Lift™/Safety-Lock™ Lid
Specifications
Rotor Adapter | |
ROTINA 380/380 R Centrifuge with Carrier no. 1752 |