Learn More
Hettich™ Rotor Adapter for Carrier no. 1425
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand: Hettich™ 1437
Additional Details : Weight : 0.01000kg
Description
- Rotor adapter for Hettich Carriers
- For use with Universal 320/320 R Centrifuge
- Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
Specifications
Adapter | |
1 x 100 mL Tube | |
100 mm | |
1 |
Round Bottom | |
44 mm | |
Carrier no 1425 and Swing-out Rotor 1494 for Universal 320/320 R Centrifuge |