Learn More
Tocris Bioscience™ Rolipram
GREENER_CHOICE
Product Code. 30001437
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 30001437
Brand: Tocris Bioscience™ 0905/10
PDE4 inhibitor
Rolipram is a selective inhibitor of cAMP phosphodiesterase (PDE4) (IC50 = 2.0 μM). Discriminates between two conformational states of PDE4 isoenzymes.
R-enantiomer also available.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 61413-54-5
Molecular Formula C16H21NO3
Specifications
Quantity 10 mg
Formula Weight 275.35
Purity >98%
Molecular Weight (g/mol) 275.35
Chemical Name or Material Rolipram