Learn More
Hamilton™ Robotics Polyplast Temp BNC Electrode
Click to view available options
Packaging Quantity:
1 Each
Unit Size:
Each
Specifications
Specifications
| Type | Polyplast Temp BNC Electrode |
| Diameter (Metric) Outer | 12 mm |
| Temperature (Metric) Max | 60°C |
| For Use With (Application) | For Portable Measurement for Basic Laboratory Applications |
| Packaging Quantity | 1 Each |
| Includes | Pt1000 Temperature sensor |
| Overall Length (Metric) | 120 mm |
| Material | Plastic (SAN), Glass |