Learn More
RNF145 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 24524-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16892995
  • 3468.15 NOK / 150µL
Estimated Shipment: 19-12-2022
Description
The RNF145 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human RNF145. This antibody recognizes human antigen. The RNF145 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
RNF145
Polyclonal
Unconjugated
150 μL
153830
IgG
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
367 μg/mL