Learn More
Rnase-free Water, non DEPCAbnova™
Click to view available options
Quantity:
5 x 1 mL
2 x 50 mL
500 mL
Description
- The RNase-free Water, non DEPC was prepared from highly purified water by ultrafiltration without DEPC treatment.
- The product was confirmed to be free from RNase and DNase.
- Apperance is clear liquid.
Specifications
Quantity | 5 x 1 mL |