Learn More
Rifapentine, MedChemExpress
Product Code. 16464317
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Product Code. 16464317
Brand: Medchem Express HYB0269
MedChemExpress Rifapentine (DL 473) is an antibiotic compound used in the treatment of tuberculosis.
Rifapentine (DL 473) is an antibiotic compound used in the treatment of tuberculosis.
Specifications
Chemical Name or Material Rifapentine
Physical Form Solid
CAS 61379-65-5
Color Red
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C47H64N4O12
For Use With (Application) COVID-19-anti-virus
Synonym DL 473 Cyclopentylrifampicin
Solubility Information DMSO : ≥ 50 mg/mL (57.01 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES OC(C1=C(C2=O)C(O[C@@]2(O/C=C/[C@@H]([C@H]([C@@]([C@@H]3C)([H])OC(C)=O)C)OC)C)=C(C)C(O)=C1C(O)=C4NC(/C(C)=C\C=C\[C@@H]([C@@H]([C@@H](C)[C@H]3O)O)C)=O)=C4/C=N/N5CCN(C6CCCC6)CC5
Molecular Weight (g/mol) 877.03
Formula Weight 877.03
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.