Learn More
Thermo Scientific Chemicals Rhodamine 110, 98+%
Catalog Number p-4509388
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
13558-31-1
C20H15ClN2O3
366.801
JNGRENQDBKMCCR-UHFFFAOYSA-N
11617572
2-(3-amino-6-iminoxanthen-9-yl)benzoic acid;hydrochloride
C1=CC=C(C(=C1)C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N)C(=O)O.Cl
13558-31-1
C20H15ClN2O3
366.801
JNGRENQDBKMCCR-UHFFFAOYSA-N
11617572
2-(3-amino-6-iminoxanthen-9-yl)benzoic acid;hydrochloride
C1=CC=C(C(=C1)C2=C3C=CC(=N)C=C3OC4=C2C=CC(=C4)N)C(=O)O.Cl
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point >360.0°C
Assay Percent Range 98+%
Fluorescence Max 520nm (in methanol)
Solubility Information Solubility in water: slightly soluble.
Infrared Spectrum Authentic
Lambda Max 498 to 504nm (c=10mg/l in methanol)
Formula Weight 366.79
Percent Purity ≥98%
Grade Pure
Molar Absorption Coefficient 77000 l/(mol.cm) (at lambda max.) min.
Packaging Glass bottle
Suitability (for use as a laser dye) passes
Color White to Yellow
Quantity 500 mg
Physical Form Crystalline Powder
Chemical Name or Material Rhodamine 110