missing translation for 'onlineSavingsMsg'
Learn More

Retene, 97%, Thermo Scientific Chemicals

Catalog Number p-7077570
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
483-65-8
C18H18
234.34
MFCD00016358
NXLOLUFNDSBYTP-UHFFFAOYSA-N
retene, reten, 1-methyl-7-isopropylphenanthrene, 7-isopropyl-1-methylphenanthrene, phenanthrene, 1-methyl-7-1-methylethyl, 1-methyl-7-1-methylethyl phenanthrene, unii-0w2d2e1p9q, phenanthrene, 7-isopropyl-1-methyl, ccris 3180, 7-isopropyl-1-methylphenanthren
10222
1-methyl-7-propan-2-ylphenanthrene
CC(C)C1=CC=C2C(C=CC3=C(C)C=CC=C23)=C1
This item is not returnable. View return policy
483-65-8
C18H18
234.34
MFCD00016358
NXLOLUFNDSBYTP-UHFFFAOYSA-N
retene, reten, 1-methyl-7-isopropylphenanthrene, 7-isopropyl-1-methylphenanthrene, phenanthrene, 1-methyl-7-1-methylethyl, 1-methyl-7-1-methylethyl phenanthrene, unii-0w2d2e1p9q, phenanthrene, 7-isopropyl-1-methyl, ccris 3180, 7-isopropyl-1-methylphenanthren
10222
1-methyl-7-propan-2-ylphenanthrene
CC(C)C1=CC=C2C(C=CC3=C(C)C=CC=C23)=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 97.0°C to 99.0°C
Boiling Point 108.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 250 mg
Solubility Information Solubility in water: negligible. Other solubilities: soluble in hot alochol, benzene, hot ether, carbon, disulfide
Formula Weight 234.34
Percent Purity 97%
Chemical Name or Material Retene
compliance-icons
Product Identifier
  • Retene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'