Learn More
Eppendorf™ Research™ plus EU-IVD Single Channel Fixed Volume Pipette
Meets the highest needs in precision and accuracy – combined with ultimate ergonomics and increased flexibility.
Brand:  Eppendorf™ 3121000058
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 10040754
  • 1838.00 NOK / Each
Estimated Shipment
15-02-2021
Add to basket
Description
Description
  • Air-displacement pipette for accurate pipetting of aqueous solutions
  • Feel the difference in weight and pipetting forces: the ultra light mechanical pipette is designed according to the strict criteria of the Eppendorf PhysioCare Concept and limits the strain on your hand and arm
  • Spring loaded tip cone (not available for 5mL and 10mL pipettes) for minimal tip attachment forces helps to reduce stress
  • Low tip ejection force of Eppendorf Research plus can be as low as 3.6 N
  • Adjust your pipette in seconds for better accuracy when pipetting various difficult liquids like ethanol or even when pipetting at high altitudes. Return to factory adjustment without calibration
Specifications
Specifications
±1%
In vitro diagnostic device according to Directive 98/79/EC of the European Parliament and the Council dated October 27, 1998
Spring loaded tip cone, ejector, yellow operating button
25 μL
Autoclavable
200μL Pipette tip