Learn More
Trajan Scientific and Medical™ Replacement Plunger for Syringes with Valves
Replacement plungers for gas tight syringes.
Brand:  Trajan Scientific and Medical™ 031842
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15208563
Quantity Price
1 585.88 NOK / Each
10 543.52 NOK / Each
Estimated Shipment
20-08-2020
Add to basket
Description
Description
  • Replacement plungers for gas tight syringes
  • Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
23 G
0.63 mm
Bevel
1 mL
0.32 mm
50 mm
Replacement Plunger for Syringes with Valves